Assay Details
| Gene Symbol: LOC102188176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676340.3 | XP_005676397.2 | 23 - 24 | 2721 | 108 | |
| XM_005676341.3 | XP_005676398.2 | 23 - 24 | 2721 | 108 | ||
| XM_005676342.3 | XP_005676399.2 | 23 - 24 | 2721 | 108 | ||
| XM_005676343.3 | XP_005676400.2 | 23 - 24 | 2721 | 108 | ||
| GenBank mRNA | KA326311.1 | - | 4 - 5 | 425 | 108 | |
More Information
Set Membership: |
|

LOC102188176
Genomic Map