Assay Details
| Gene Symbol: ILF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677489.3 | XP_005677546.1 | 7 - 8 | 597 | 71 | |
| GenBank mRNA | JO388133.2 | - | 7 - 8 | 1112 | 71 | |
| JO579532.1 | - | 7 - 8 | 580 | 71 | ||
More Information
Set Membership: |
|

ILF2
Genomic Map