Assay Details
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677757.3 | XP_005677814.2 | 9 - 10 | 1611 | 62 | |
| GenBank mRNA | JT645807.1 | - | 9 - 10 | 995 | 62 | |
More Information
Set Membership: |
|

ACP6
Genomic Map