Assay Details
| Gene Symbol: KCNA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677998.3 | XP_005678055.3 | 1 - 2 | 3497 | 96 | |
| GenBank mRNA | JT645988.1 | - | 3 - 4 | 291 | 96 | |
More Information
Set Membership: |
|

KCNA3
Genomic Map