Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678156.3 | XP_005678213.2 | 5 - 6 | 862 | 65 | |
| GenBank mRNA | JO645667.2 | - | 1 - 2 | 3031 | 65 | |
More Information
Set Membership: |
|

DDAH1
Genomic Map