Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678554.2 | XP_005678611.1 | 3 - 4 | 378 | 65 | |
| GenBank mRNA | JO605881.2 | - | 18 - 19 | 2377 | 65 | |
| KA322284.1 | - | 3 - 4 | 320 | 65 | ||
More Information
Set Membership: |
|

KIF2C
Genomic Map