Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047340.1 | XP_017902829.1 | 2 - 3 | 232 | 66 | |
| XM_018047341.1 | XP_017902830.1 | 2 - 3 | 231 | 66 | ||
| XM_018047342.1 | XP_017902831.1 | 2 - 3 | 295 | 66 | ||
| XM_018047343.1 | XP_017902832.1 | 3 - 4 | 371 | 66 | ||
| XM_018047344.1 | XP_017902833.1 | 3 - 4 | 388 | 66 | ||
More Information
Set Membership: |
|

MACC1
Genomic Map