Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047341.1 | XP_017902830.1 | 3 - 4 | 2271 | 86 | |
| GenBank mRNA | JT640332.1 | - | 2 - 3 | 227 | 86 | |
More Information
Set Membership: |
|

MACC1
Genomic Map