Assay Details
| Gene Symbol: ATP5F1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682906.3 | XP_005682963.1 | 1 - 2 | 275 | 67 | |
| GenBank mRNA | JT763352.1 | - | 1 - 2 | 451 | 67 | |
| KA340689.1 | - | 1 - 2 | 277 | 67 | ||
More Information
Set Membership: |
|

ATP5F1D
Genomic Map