Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050027.1 | XP_017905516.1 | 3 - 4 | 353 | 69 | |
| GenBank mRNA | JT760224.1 | - | 2 - 3 | 201 | 69 | |
More Information
Set Membership: |
|

RFESD
Genomic Map