Assay Details
| Gene Symbol: ALKAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058127.1 | XP_017913616.1 | 3 - 4 | 311 | 113 | |
| XM_018058128.1 | XP_017913617.1 | 3 - 4 | 311 | 113 | ||
More Information
Set Membership: |
|

ALKAL1
Genomic Map