Assay Details
| Gene Symbol: PLSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064574.1 | XP_017920063.1 | 5 - 6 | 1459 | 53 | |
| XM_018064575.1 | XP_017920064.1 | 6 - 7 | 1247 | 53 | ||
| GenBank mRNA | JO578861.1 | - | 5 - 6 | 1234 | 53 | |
More Information
Set Membership: |
|

PLSCR3
Genomic Map