Assay Details
| Gene Symbol: GSDMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013972351.2 | XP_013827805.2 | 8 - 9 | 1424 | 95 | |
| GenBank mRNA | JO407878.2 | - | 4 - 5 | 639 | 95 | |
| KT983435.1 | - | 7 - 8 | 815 | 95 | ||
More Information
Set Membership: |
|

GSDMB
Genomic Map