Assay Details
| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693855.3 | XP_005693912.3 | 7 - 8 | 1514 | 62 | |
| GenBank mRNA | JO429713.2 | - | 3 - 4 | 628 | 62 | |
More Information
Set Membership: |
|

COASY
Genomic Map