Assay Details
| Gene Symbol: CAMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695630.3 | XP_005695687.2 | 5 - 6 | 548 | 68 | |
| XM_018038316.1 | XP_017893805.1 | 5 - 6 | 952 | 68 | ||
| XM_018038317.1 | XP_017893806.1 | 5 - 6 | 483 | 68 | ||
| GenBank mRNA | JT673901.1 | - | 6 - 7 | 809 | 68 | |
More Information
Set Membership: |
|

CAMK1
Genomic Map