Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695649.3 | XP_005695706.1 | 7 - 8 | 1782 | 61 | |
XM_005695650.3 | XP_005695707.1 | 7 - 8 | 1782 | 61 | ||
GenBank mRNA | JO622928.2 | - | 4 - 5 | 1724 | 61 | |
JO622929.2 | - | 3 - 4 | 1632 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
