Assay Details
| Gene Symbol: LOC102188999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697765.3 | XP_005697822.2 | 13 - 14 | 1855 | 96 | |
| GenBank mRNA | JO426248.1 | - | 13 - 14 | 1929 | 96 | |
| JO428960.2 | - | 13 - 14 | 1869 | 96 | ||
More Information
Set Membership: |
|

LOC102188999
Genomic Map