Assay Details
| Gene Symbol: ZNF250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058795.1 | XP_017914284.1 | 3 - 4 | 287 | 60 | |
| XM_018058796.1 | XP_017914285.1 | 3 - 4 | 1422 | 60 | ||
| XM_018058797.1 | XP_017914286.1 | 3 - 4 | 994 | 60 | ||
| GenBank mRNA | JO448848.2 | - | 1 - 2 | 216 | 60 | |
More Information
Set Membership: |
|

ZNF250
Genomic Map