Assay Details
| Gene Symbol: DTX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059025.1 | XP_017914514.1 | 8 - 9 | 1662 | 57 | |
| XM_018059026.1 | XP_017914515.1 | 8 - 9 | 2324 | 57 | ||
| XM_018059027.1 | XP_017914516.1 | 8 - 9 | 2050 | 57 | ||
More Information
Set Membership: |
|

DTX4
Genomic Map