Assay Details
| Gene Symbol: KMT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061359.1 | XP_017916848.1 | 4 - 5 | 593 | 84 | |
| XM_018061360.1 | XP_017916849.1 | 5 - 6 | 670 | 84 | ||
| XM_018061361.1 | XP_017916850.1 | 5 - 6 | 775 | 84 | ||
| XM_018061362.1 | XP_017916851.1 | 4 - 5 | 973 | 84 | ||
More Information
Set Membership: |
|

KMT5A
Genomic Map