Assay Details
| Gene Symbol: KMT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061359.1 | XP_017916848.1 | 6 - 7 | 837 | 72 | |
| XM_018061360.1 | XP_017916849.1 | 7 - 8 | 914 | 72 | ||
| XM_018061361.1 | XP_017916850.1 | 7 - 8 | 1019 | 72 | ||
| XM_018061362.1 | XP_017916851.1 | 6 - 7 | 1217 | 72 | ||
More Information
Set Membership: |
|

KMT5A
Genomic Map