Assay Details
| Gene Symbol: UFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044839.1 | XP_017900328.1 | 5 - 6 | 972 | 64 | |
| XM_018044840.1 | XP_017900329.1 | 5 - 6 | 487 | 64 | ||
More Information
Set Membership: |
|

UFD1
Genomic Map