Assay Details
Gene Symbol: KRTDAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971229.2 | XP_013826683.1 | 5 - 6 | 309 | 62 | |
XM_018063392.1 | XP_017918881.1 | 4 - 5 | 267 | 62 | ||
GenBank mRNA | KA338285.1 | - | 467 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
