Assay Details
| Gene Symbol: GRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062705.1 | XP_017918194.1 | 1 - 2 | 522 | 61 | |
| GenBank mRNA | JO618160.2 | - | 1 - 2 | 240 | 61 | |
| KA315529.1 | - | 2 - 3 | 204 | 61 | ||
More Information
Set Membership: |
|

GRWD1
Genomic Map