Assay Details
| Gene Symbol: ILRUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038991.1 | XP_017894480.1 | 3 - 4 | 748 | 70 | |
| XM_018038993.1 | XP_017894482.1 | 3 - 4 | 704 | 70 | ||
More Information
Set Membership: |
|

ILRUN
Genomic Map