Assay Details
| Gene Symbol: N6AMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674721.3 | XP_005674778.1 | 2 - 3 | 244 | 54 | |
| GenBank mRNA | JO456523.2 | - | 3 - 4 | 314 | 54 | |
| JT705990.1 | - | 4 - 5 | 413 | 54 | ||
| KA350561.1 | - | 2 - 3 | 241 | 54 | ||
More Information
Set Membership: |
|

N6AMT1
Genomic Map