Assay Details
| Gene Symbol: NAALADL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675296.3 | XP_005675353.2 | 9 - 10 | 2126 | 77 | |
| XM_013964525.2 | XP_013819979.2 | 11 - 12 | 1880 | 77 | ||
| GenBank mRNA | KA311265.1 | - | 1 - 2 | 133 | 77 | |
More Information
Set Membership: |
|

NAALADL2
Genomic Map