Assay Details
| Gene Symbol: AMMECR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065881.1 | XP_017921370.1 | 3 - 4 | 852 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMMECR1L| Gene Symbol: AMMECR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065881.1 | XP_017921370.1 | 3 - 4 | 852 | 60 | |
Set Membership: |
|