Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679206.3 | XP_005679263.2 | 6 - 7 | 634 | 101 | |
| XM_018047129.1 | XP_017902618.1 | 7 - 8 | 739 | 101 | ||
| XM_018047131.1 | XP_017902620.1 | 7 - 8 | 591 | 101 | ||
| XM_018047132.1 | XP_017902621.1 | 7 - 8 | 520 | 101 | ||
| XM_018047133.1 | XP_017902622.1 | 6 - 7 | 416 | 101 | ||
| GenBank mRNA | JT713066.1 | - | 6 - 7 | 495 | 101 | |
| KA313758.1 | - | 7 - 8 | 678 | 101 | ||
More Information
Set Membership: |
|

NT5C3A
Genomic Map