Assay Details
| Gene Symbol: POC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047611.1 | XP_017903100.1 | 11 - 12 | 3122 | 68 | |
| XM_018047612.1 | XP_017903101.1 | 10 - 11 | 1373 | 68 | ||
More Information
Set Membership: |
|

POC1B
Genomic Map