Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680371.3 | XP_005680428.1 | 4 - 5 | 593 | 61 | |
| XM_005680372.3 | XP_005680429.1 | 4 - 5 | 575 | 61 | ||
More Information
Set Membership: |
|

PA2G4
Genomic Map