Assay Details
| Gene Symbol: GPAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686247.3 | XP_005686304.3 | 11 - 12 | 1521 | 67 | |
| XM_018054835.1 | XP_017910324.1 | 5 - 6 | 808 | 67 | ||
More Information
Set Membership: |
|

GPAT2
Genomic Map