Assay Details
| Gene Symbol: BABAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686868.3 | XP_005686925.1 | 4 - 5 | 556 | 84 | |
| GenBank mRNA | JO431565.2 | - | 4 - 5 | 442 | 84 | |
| KA340956.1 | - | 4 - 5 | 451 | 84 | ||
More Information
Set Membership: |
|

BABAM2
Genomic Map