Assay Details
| Gene Symbol: YWHAQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055677.1 | XP_017911166.1 | 2 - 3 | 508 | 79 | |
| XM_018055678.1 | XP_017911167.1 | 2 - 3 | 434 | 79 | ||
| GenBank mRNA | JO580719.1 | - | 1 - 2 | 329 | 79 | |
| JO631704.2 | - | 1 - 2 | 778 | 79 | ||
More Information
Set Membership: |
|

YWHAQ
Genomic Map