Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687955.3 | XP_005688012.1 | 4 - 5 | 412 | 82 | |
| XM_005687956.3 | XP_005688013.1 | 3 - 4 | 385 | 82 | ||
| XM_018057128.1 | XP_017912617.1 | 3 - 4 | 270 | 82 | ||
More Information
Set Membership: |
|

PRPF18
Genomic Map