Assay Details
| Gene Symbol: RRM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058322.1 | XP_017913811.1 | 3 - 4 | 538 | 73 | |
| GenBank mRNA | JT697416.1 | - | 2 - 3 | 440 | 73 | |
More Information
Set Membership: |
|

RRM2B
Genomic Map