Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061973.1 | XP_017917462.1 | 7 - 8 | 1043 | 78 | |
| GenBank mRNA | KA334242.1 | - | 9 - 10 | 1220 | 78 | |
More Information
Set Membership: |
|

DNAJA2
Genomic Map