Assay Details
| Gene Symbol: SCN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063385.1 | XP_017918874.1 | 4 - 5 | 927 | 69 | |
| GenBank mRNA | JT702623.1 | - | 5 - 6 | 659 | 69 | |
More Information
Set Membership: |
|

SCN1B
Genomic Map