Assay Details
| Gene Symbol: TRIM50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040552.1 | XP_017896041.1 | 3 - 4 | 761 | 67 | |
| XM_018040553.1 | XP_017896042.1 | 3 - 4 | 761 | 67 | ||
More Information
Set Membership: |
|

TRIM50
Genomic Map