Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042516.1 | XP_017898005.1 | 5 - 6 | 1019 | 86 | |
| XM_018042517.1 | XP_017898006.1 | 4 - 5 | 812 | 86 | ||
| XM_018042519.1 | XP_017898008.1 | 5 - 6 | 989 | 86 | ||
| XM_018042520.1 | XP_017898009.1 | 5 - 6 | 988 | 86 | ||
| XM_018042521.1 | XP_017898010.1 | 5 - 6 | 1380 | 86 | ||
| XM_018042522.1 | XP_017898011.1 | 2 - 3 | 586 | 86 | ||
| GenBank mRNA | KA329347.1 | - | 1 - 2 | 1650 | 86 | |
More Information
Set Membership: |
|

CHST3
Genomic Map