Assay Details
| Gene Symbol: LOC108633235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043551.1 | XP_017899040.1 | 7 - 8 | 908 | 104 | |
| Gene Symbol: LOC102175469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043600.1 | XP_017899089.1 | 7 - 8 | 908 | 104 | |
More Information
Set Membership: |
|

LOC108633235
Genomic Map