Assay Details
| Gene Symbol: SLC39A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048082.1 | XP_017903571.1 | 8 - 9 | 1378 | 64 | |
| XM_018048083.1 | XP_017903572.1 | 8 - 9 | 1711 | 64 | ||
| XM_018048084.1 | XP_017903573.1 | 8 - 9 | 1710 | 64 | ||
| GenBank mRNA | JO614432.2 | - | 3 - 4 | 345 | 64 | |
| JP351464.2 | - | 3 - 4 | 535 | 64 | ||
| JT666826.1 | - | 7 - 8 | 3500 | 64 | ||
More Information
Set Membership: |
|

SLC39A5
Genomic Map