Assay Details
| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049182.1 | XP_017904671.1 | 5 - 6 | 1215 | 61 | |
| XM_018049183.1 | XP_017904672.1 | 5 - 6 | 1215 | 61 | ||
More Information
Set Membership: |
|

PRSS12
Genomic Map