Assay Details
| Gene Symbol: TRIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057667.1 | XP_017913156.1 | 2 - 3 | 554 | 83 | |
| GenBank mRNA | JO419327.2 | - | 2 - 3 | 439 | 83 | |
| JT682324.1 | - | 3 - 4 | 1714 | 83 | ||
More Information
Set Membership: |
|

TRIB3
Genomic Map