Assay Details
| Gene Symbol: CCKBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059455.1 | XP_017914944.1 | 2 - 3 | 502 | 56 | |
| XM_018059456.1 | XP_017914945.1 | 2 - 3 | 691 | 56 | ||
| XM_018059457.1 | XP_017914946.1 | 3 - 4 | 397 | 56 | ||
| GenBank mRNA | KC767906.1 | - | 1 - 2 | 151 | 56 | |
More Information
Set Membership: |
|

CCKBR
Genomic Map