Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063156.1 | XP_017918645.1 | 2 - 3 | 1176 | 125 | |
| XM_018063157.1 | XP_017918646.1 | 2 - 3 | 1180 | 125 | ||
| GenBank mRNA | JO440828.2 | - | 2 - 3 | 488 | 125 | |
| KA346148.1 | - | 2 - 3 | 1042 | 125 | ||
More Information
Set Membership: |
|

ARL6IP6
Genomic Map