Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063156.1 | XP_017918645.1 | 3 - 4 | 1313 | 72 | |
| GenBank mRNA | JO440828.2 | - | 3 - 4 | 625 | 72 | |
| KA346148.1 | - | 3 - 4 | 1179 | 72 | ||
More Information
Set Membership: |
|

ARL6IP6
Genomic Map