Assay Details
| Gene Symbol: KIAA2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060816.1 | XP_017916305.1 | 21 - 22 | 3508 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA2012| Gene Symbol: KIAA2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060816.1 | XP_017916305.1 | 21 - 22 | 3508 | 56 | |
Set Membership: |
|