Assay Details
| Gene Symbol: DNAAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063092.1 | XP_017918581.1 | 1 - 2 | 1042 | 58 | |
| XM_018063093.1 | XP_017918582.1 | 1 - 2 | 1042 | 58 | ||
| XM_018063095.1 | XP_017918584.1 | 1 - 2 | 1042 | 58 | ||
| XM_018063096.1 | XP_017918585.1 | 1 - 2 | 1042 | 58 | ||
| XM_018063097.1 | XP_017918586.1 | 5 - 6 | 680 | 58 | ||
| XM_018063098.1 | XP_017918587.1 | 1 - 2 | 1042 | 58 | ||
| XM_018063099.1 | XP_017918588.1 | 1 - 2 | 1042 | 58 | ||
More Information
Set Membership: |
|

DNAAF3
Genomic Map