Assay Details
| Gene Symbol: RFFL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064231.1 | XP_017919720.1 | 4 - 5 | 883 | 94 | |
| XM_018064232.1 | XP_017919721.1 | 4 - 5 | 1329 | 94 | ||
| XM_018064233.1 | XP_017919722.1 | 4 - 5 | 880 | 94 | ||
| GenBank mRNA | KA316003.1 | - | 3 - 4 | 364 | 94 | |
More Information
Set Membership: |
|

RFFL
Genomic Map